Learn More
Thermo Scientific™ Tiras G M.I.C.Evaluator (M.I.C.E.™) con penicilina
Determine los valores exactos de concentración inhibitoria mínima (CIM) de los antibióticos utilizando las tiras reactivas MIC.
Marca:  Thermo Scientific™ MA0101D
Detalles adicionales : Peso : 0.04000kg
 Ver más versiones de este producto
Código de producto. 11494009
  • 46.86€ / paquete de 10
Fecha de envío estimada
del proveedor 30-01-2019
Añadir a la cesta
Descripción
Descripción
Conveniently packaged M.I.C.E. strips combine the ease of a diffusion test with the accuracy of an MIC test.
Simple to Perform
  • Distinctive gradient format provides excellent contrast with the agar
  • Increased font size makes reading easier
  • Scale conforms to international standards
  • Strips are tailored to meet clinical requirements while maintaining the accuracy required for research
Easy-to-handle
  • The strip is conveniently presented when the sachet is peeled apart
  • Single-strip packaging eliminates the requirement to maintain and quality control opened strips; open one and use it
  • Stability of the individual M.I.C.E. strips is ensured through the inclusion of a desiccant within each sachet
  • 10 and 50 pack sizes offer users maximum flexibility to put together custom panels for testing
  • Easily stackable boxes simplify storage; all antimicrobials are stored in the fridge (2-8°C)
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Especificación
Especificación
P
10 Strips/Pk.
256-0.015µg/mL
Penicillin G M.I.C.Evaluator Strips (M.I.C.E.™)
2-8°C
Videos