Learn More
3M™ Casco de protección Peltor™ G3000
Series G3000 Ventilated Ratchet Hard Hat allows falling objects to slide off easily and not dislodge the helmet from its position.
Marca: 3M™ 7100001960
19.70 EUR hasta el 2024-12-31
Use el código promocional "22912" para aplicar la oferta.
Detalles adicionales : Peso : 0.36000kg
Descripción
- A hard hat designed for use in harsh environments, with excellent ventilation and a good field of vision.
- 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV- radiation and its time to replace it.
- Softly rounded design, which helps to avoid helmet parts snagging on branches, etc.
- Ventilation holes distributed over a larger area offer maximum through-flow of air increasing wearer comfort.
- Head harness can be rotated 180° for better visibility when working in confined spaces.
- Short peak gives improved upwards visibility.
Especificaciones
EN397 | |
ABS estabilizado ante UV | |
Casco de protección | |
Sí |
Blanco | |
G30NUW | |
1 unidad |