Learn More
PDPR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA561648
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: LEKGMDFIGR DALLQQKQNG VYKRLTMFIL DDHDSDLDLW PWWGEPIYRN GQYVGKTTSS AYSYSLERHV CLGFVHNFSE DTGEEQVVTA DFINRG Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.
Decreases the sensitivity of PDP1 to magnesium ions, and this inhibition is reversed by the polyamine spermine.Especificaciones
PDPR | |
Polyclonal | |
Unconjugated | |
PDPR | |
4930402E16Rik; AU018056; Kiaa1990; mKIAA1990; PDP3; PDPR; pyruvate dehydrogenase phosphatase regulatory subunit; pyruvate dehydrogenase phosphatase regulatory subunit, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55066 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NCN5 | |
PDPR | |
Recombinant protein corresponding to Human PDPR | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only