missing translation for 'onlineSavingsMsg'
Learn More

PDILT Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP2-83378-0.1ML

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213749

  • 455.71€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

PDILT Polyclonal specifically detects PDILT in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

PDILT
Polyclonal
PBS, 2% Sucrose
Rabbit
Immunogen affinity purified
RUO
204474
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
PDIA7, protein disulfide isomerase family A, member 7, protein disulfide isomerase-like protein of the testis, protein disulfide isomerase-like, testis expressed, protein disulfide-isomerase-like protein of the testis
The immunogen is a synthetic peptide directed towards the C-terminal region of Human PDILT
0.1 mL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales