missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PDIA3 (Human) Recombinant Protein
Human PDIA3 full-length ORF ( NP_005304.3, 1 a.a. - 505 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00002923-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MRLRRLALFPGVALLLAAARLAAASDVLELTDDNFESRISDTGSAGLMLVEFFAPWCGHCKRLAPEYEAAATRLKGIVPLAKVDCTANTNTCNKYGVSGYPTLKIFRDGEEAGAYDGPRTADGIVSHLKKQAGPASVPLRTEEEFKKFISDKDASIVGFFDDSFSEAHSEFLKAASNLRDNYRFAHTNVESLVNEYDDNGEGIILFRPSHLTNKFEDKTVAYTEQKMTSGKIKKFIQENIFGICPHMTEDNKDLIQGKDLLIAYYDVDYEKNAKGSNYWRNRVMMVAKKFLDAGHKLNFAVASRKTFSHELSDFGLESTAGEIPVVAIRTAKGEKFVMQEEFSRDGKALERFLQDYFDGNLKRYLKSEPIPESNDGPVKVVVAENFDEIVNNENKDVLIEFYAPWCGHCKNLEPKYKELGEKLSKDPNIVIAKMDATANDVPSPYEVRGFPTIYFSPANKKLNPKKYEGGRELSDFISYLQREATNPPVIQEEKPKKKKKAQEDL
Especificaciones
NP_005304.3 | |
protein disulfide isomerase family A, member 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PDIA3 | |
GST |
2923 | |
Wheat germ expression system | |
25 μg | |
ER60, ERp57, ERp60, ERp61, GRP57, GRP58, HsT17083, P58, PI-PLC | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |