missing translation for 'onlineSavingsMsg'
Learn More

PDGFR alpha Antibody (RM0004-3G28), DyLight 405, Novus Biologicals™

Rat Monoclonal Antibody

Marca:  Bio-Techne NB110-61020V

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216173

  • 590.70€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

PDGFR alpha Monoclonal specifically detects PDGFR alpha in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

PDGFR alpha
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
PDGFRA
Mouse PDGFR alpha (extracellular domain)
0.1 mL
Phospho Specific, Protein Kinase
5156
Mouse
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
RM0004-3G28
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA
Rat
Protein G purified
RUO
Primary
This antibody recognizes mouse PDGF receptor alpha in western blot. No cross reactivity to mouse PDGF receptor beta.
Store at 4C in the dark.
IgG2
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only