missing translation for 'onlineSavingsMsg'
Learn More

PDGF-C Antibody (004), DyLight 650, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-89373C

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215129

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

PDGF-C Monoclonal specifically detects PDGF-C in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

PDGF-C
Monoclonal
DyLight 650
FALLOTEIN, PDGF-C, platelet derived growth factor C, platelet-derived growth factor C, SCDGFSpinal cord-derived growth factor, secretory growth factor-like protein, VEGF-E
This antibody was obtained from a rabbit immunized with purified, recombinant Human PDGF-C (NP_057289.1; Val235-Gly345).
0.1 mL
Cell Cycle and Replication
56034
Store at 4°C in the dark.
IgG
ELISA, Immunofluorescence
4
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales