missing translation for 'onlineSavingsMsg'
Learn More

PDDC1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP2-83374-0.1ML

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214059

  • 455.71€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

PDDC1 Polyclonal specifically detects PDDC1 in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

PDDC1
Polyclonal
PBS, 2% Sucrose
Rabbit
Immunogen affinity purified
RUO
347862
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
FLJ35497, Parkinson disease 7 domain containing 1, Parkinson disease 7 domain-containing protein 1
The immunogen is a synthetic peptide directed towards the middle region of human PDDC1
0.1 mL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales