missing translation for 'onlineSavingsMsg'
Learn More

PCDH1 (Protocadherin 1) Mouse anti-Human, Clone: CSTEM31, eBioscience™

Mouse Monoclonal Antibody

Marca:  Invitrogen 14-9157-80

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923194

  • 113.00€ / 25 microgramos


Fecha estimada de envío
20-08-2020
Añadir a la cesta

Descripción

Descripción

Protocadherin 1, also known as PCDH1 and cadherin-like protein 1, is a transmembrane protein involved in cell-cell adhesion. Protocadherin 1 belongs to a group of cadherin-like proteins termed delta1-protocadherins, which consists of protocadherin-1, -7, -9, and -11(X/Y). The delta1-protocadherins have seven extracellular cadherin-repeats, a transmembrane domain, and a cytoplasmic domain, and can be expressed in two isoforms, short or long. Expression of protocadherin 1 is regulated both temporally and spatially, and plays a role both in early development and mature tissues. In mice, it is expressed in a number of tissues and organs, including the brain, spinal cord, peripheral ganglia, inner ear, hair follicles, kidney, reproductive organs, prostate, and the seminal gland. Notably, protocadherin 1 is a susceptibility gene for Bronchial Hyperresponsiveness in humans. Expression of protocadherin 1 has also been identified on human pluripotent stem cells.
Especificaciones

Especificaciones

PCDH1 (Protocadherin 1)
Monoclonal
0.5 mg/mL
PBS with proprietary stabilizer and 0.09% sodium azide; pH 7.2
Q08174
PCDH1, cadherin-like protein 1, protocadherin-42
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence
CSTEM31
Unconjugated
PCDH1
Liquid
PCDH1
IgG2b, kappa
25 μg
4° C, do not freeze
Primary
5097
Videos
Documentos