missing translation for 'onlineSavingsMsg'
Learn More

PBEF/Visfatin/NAMPT Antibody (OMNI379), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-80036R

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214497

  • 673.85€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

PBEF/Visfatin/NAMPT Monoclonal antibody specifically detects PBEF/Visfatin/NAMPT in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Especificaciones

Especificaciones

PBEF/Visfatin/NAMPT
Monoclonal
DyLight 550
EC 2.4.2.12, MGC117256, NAmPRTase, Nampt, nicotinamide phosphoribosyltransferase, PBEF1110035O14Rik, PBEF1DKFZp666B131, Pre-B cell-enhancing factor, pre-B-cell colony enhancing factor 1, Pre-B-cell colony-enhancing factor 1, VF, VISFATIN
Recombinant human PBEF/Visfatin/NAMPT (visfatin/PBEF).
0.1 mL
Adaptive Immunity, Cell Cycle and Replication, Immunology, Innate Immunity
10135
Store at 4°C in the dark.
IgG2a κ
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
OMNI379
50mM Sodium Borate
Mouse
Protein G purified
RUO
Primary
Human, Mouse, Rat
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales