missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PB1 Recombinant Protein

en la oferta

Especificaciones

Reactividad cruzada Human
Formulación 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Peso molecular 59.4
Nombre PBRM1 (Human) Recombinant Protein (P01)
Intervalo de pH 8
Ver más especificaciones

Descripción

Descripción

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Especificaciones

Especificaciones

Human
59.4
8
Glutathione Sepharose 4 Fast Flow
Yes
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
PBRM1 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Videos
SDS
Documentos

Documentos

Product Certifications