Learn More
PAX8 Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech CL594-10336-100UL
Detalles adicionales : Peso : 0.00010kg
Código de producto. 17254766
  • 367.00€ / 100 microlitros
Fecha estimada de envío: 13-05-2024
para ver el stock
Descripción
Descripción
PAX8 is a member of the paired box (PAX) family of transcription factors, typically containing a paired box domain and a paired-type homeodomain. It is expressed during organogenesis of the thyroid gland, kidney and Mullerian system. It is thought to regulate the expression of Wilms tumor suppressor (WT1) gene and mutations in PAX8 have been associated with Wilms tumor cells, thyroid and ovarian carcinomas. PAX8 is a useful marker in distinguishing ovarian carcinomas from mammary carcinomas (PMID: 18724243). PAX8 is expressed in a high percentage of ovarian serous, endometrioid, and clear cell carcinomas, but only rarely in primary ovarian mucinous adenocarcinomas. This antibody can detect all 5 isoforms of PAX8 (31 kDa, 34 kDa, 41 kDa, 43 kDa and 48 kDa). A 58-60 kDa form has also been reported (PMID:15650356, 1049952). Proteintech's 10336-1-AP antibody has cross-reactivities with the other PAX family members.
Especificaciones
Especificaciones
PAX8
Polyclonal
CoraLite 594
100 μL
7849
Liquid
Immunofluorescence
1000 μg/mL
Rabbit
Primary
Human, Mouse