Learn More
Emma Safety Footwear Paul XD Safety Shoes
Product Code. 15968861
Change view
Click to view available options
Size:
37
38
47
48
Unit Size:
1 pair
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15968861 48 1 pair
15938861 37 1 pair
15948861 38 1 pair
15958861 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15968861 Supplier Emma Safety Footwear Supplier No. MM70186648
  • ONLINE EXCLUSIVE PRICE
    143.14€
    83.95€ / 1 pares
    Ahorro 59.19€
    41% Descuento
Fecha estimada de envoi: 31-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
An ideal choice when optimal foot protection is required. A strong, solid steel plate in the sole and resilient leather uppers defend the feet from unexpected hazards. Perfectly balanced, anatomical arch support will keep feet feeling fit and fresh.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Design Paul XD
Material Leather
Size 48