Learn More
Applied Biosystems™ Path-ID™ qPCR Master Mix
Product Code. 30057778 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
30057778 500 Reactions 500 reactions
30057772 100 Reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30057778 Supplier Applied Biosystems™ Supplier No. 4388644
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Ambion™ Path-ID™ qPCR Master Mix is a convenient enzyme, buffer, and dNTP mix for TaqMan™ probe-based, real-time PCR detection of DNA targets.
Ambion™ Path-ID™ qPCR Master Mix is optimized for targets of importance in animal health and to function in the presence of PCR inhibitors. The mix is supplied in one tube suitable for 100 reactions. With the exception of PCR primers and probes for your target, the master mix contains everything you need for real-time PCR, including:
  • AmpliTaq Gold™ ultrapure thermostable DNA polymerase designed for automatic hot start, to minimize nonspecific PCR products
  • Optimized buffer and dNTP concentration, for reliable PCR
  • Passive internal reference, based on proprietary ROX™ dye, for precise data analysis.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2X qPCR Master Mix is stored at –20°C. Nuclease-free water may be stored at any temperature.
Detection Method Primer-probe
Form Liquid
Format Tube
PCR Method qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
Passive Reference Dye ROX (Pre-mixed)
Product Line Ambion, Path-ID
Purity or Quality Grade UP (Ultra Pure)
Quantity 500 reactions
Shipping Condition Dry Ice
For Use With (Application) Animal Health, Pathogen Detection
Hot Start Built-In Hot Start
No. of Reactions 500 Reactions
Sample Type DNA (Genomic), cDNA
Show More Show Less
For Veterinary Use Only.