missing translation for 'onlineSavingsMsg'
Learn More

Patched 1/PTCH Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications

Marca:  Bio-Techne NBP1-71662SS

Detalles adicionales : Peso : 0.00970kg

 Ver más versiones de este producto

Código de producto. 18213414

  • 185.74€ / 25 microlitros
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Patched 1/PTCH Polyclonal specifically detects Patched 1/PTCH in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Patched 1/PTCH
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
BCNSFLJ42602, FLJ26746, HPE7, NBCCS, patched (Drosophila) homolog, patched 1, patched homolog (Drosophila), patched homolog 1, patched homolog 1 (Drosophila), protein patched homolog 1, PTC, PTC1, PTCH, PTCH protein +12b, PTCH protein +4', PTCH protein -10, PTCH11
Rabbit
Affinity Purified
RUO
Primary
Human, Mouse
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
1.22 mg/mL
Western Blot 1:2500, Immunohistochemistry 1:50 - 1:250, Immunohistochemistry-Paraffin 1:50 - 1:250
Q13635
PTCH1
A genomic peptide made to a C-terminal region of the human Patched 1 protein (within residues 1200-1350). [Swiss-Prot Q13635]
0.025 mL
Cell Cycle and Replication, Stem Cell Signaling Pathway
5727
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only