missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PASK Recombinant Human Protein
PASK Recombinant Human Protein
Marca: Invitrogen PV3972
Detalles adicionales : Peso : 0.05250kg
Descripción
PASK is an evolutionarily conserved protein present in yeast, flies, and mammals. PAS domains regulate the function of many intracellular signaling pathways in response to both extrinsic and intrinsic stimuli.Especificaciones
NP_055963 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
77.2kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
PASK | |
PASK was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 533μg/mL S6 peptide substrate (AKRRRLSSLRA) and trace[32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 879-1323 | |
500nmoles of phosphate transferred to S6 peptide substrate (AKRRRLSSLRA) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 3.33μg/mL | |
GST Tag | |
Kinase and inhibitors | |
75% by SDS-PAGE |
0.34mg/mL | |
23178, 4790 | |
PASK Recombinant Human Protein | |
–80°C | |
1 Each | |
PASK Recombinant Human Protein | |
CAMKL | |
PASK Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
For Research Use Only. Not for use in diagnostic procedures.