missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PON1, Recombinant Protein

Recombinant Protein

Marca:  Invitrogen™ RP75675

Detalles adicionales : CAS : 69-74-9 Peso : 0.01000kg

Código de producto. 11598991

  • 798.00€ / 100 microgramos
Fecha estimada de envío: 23-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Paraoxonase 1 (human) Recombinant Protein, RP-75675, from Invitrogen.

Paraoxonase 1 is involved in the detoxification of organophosphate insecticides such as parathion. Paraoxonase 1 may also confer protection against coronary artery disease by destroying proinflammatory oxidized lipids present in oxidized low-density lipoproteins (LDLs). This product contains purified recombinant human Paraoxonase 1 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays. This protein has a molecular weight of ∽42.9 kDa. Purity >95% by SDS-PAGE.
Especificaciones

Especificaciones

P27169
Control, Western Blot
5444
100μg
-20°C, Avoid Freeze/Thaw Cycles
Human PON1
Human
Hu
Recombinant
Recombinant
0.12mg/mL
20mM tris HCl with 50% glycerol and no preservative
Human PON1
E. Coli
RUO
PON1
Unconjugated
E. coli
Liquid
purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales