Learn More
Titan Yellow Solution 100 mL, Ph Eur Ref 1090902, Fisher Chemical™
BIOPHARMA
Product Code. 12900564 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity Unit Size
12900564 100 mL 100mL
1 options
This item is not returnable. View return policy
Product Code. 12900564 Supplier Fisher Chemical Supplier No. RP 222 100
  • ONLINE EXCLUSIVE PRICE
    68.00€
    52.08€ / 100 ml
    Ahorro 15.92€
    23% Descuento
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Packaging Bottle
Analyte or Component Names Paper Titan Paper
Purity Grade Notes EU Ph Ref: 1090902
Quantity 100 mL
Grade Eu. Ph.