Learn More
Brady™ Paper Thermal Transfer Printable Labels
Product Code. 12683785 Shop All Brady Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 3000
Product Code. Color Unit Size
12683785 White Pack of 3000
1 options
This item is not returnable. View return policy
Product Code. 12683785 Supplier Brady™ Supplier No. 035655
  • ONLINE EXCLUSIVE PRICE
    138.00€
    114.91€ / paquete de 3000
    Ahorro 23.09€
    16% Descuento
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
B-408 Labels are thermal transfer printable and is designed to be removable after 30 days from cardboard, stainless steel, glass and polypropylene.
  • Ideal for use in general labeling and barcoding applications where the label needs to be removed or repositionable.
  • Designed to be removable after 30 days from cardboard, stainless steel, glass and polypropylene when used at ambient conditions.
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC.
  • A ribbon is required to print this material.
  • Use Part Number IP-R6100 for the BradyPrinter i5100 or BRADY IP™ Series Label Printer; use R6100 for a different printer.
Specifications
Certifications/Compliance RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Adhesive Type Acrylic, Removable
Coating None
Color White
Display Blank
Finish Matte
Format Die-cut
Material Paper
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) General identification
For Use With (Equipment) BBP81, i5100, i6100, i7100, i7500, PR Plus, IP Series, THT 76 mm Core Printers
Labels per Row 1
No. per Pack 3000 Label(s) / Roll
Quantity 1 Roll
Shape Rectangle
Size (English) 1 in H x 2 in W
Size (Metric) 25.40 mm H x 50.80 mm W
Thickness (English) 0.003 in.
Thickness (Metric) 0.086 mm
Vertical Repeat (Metric) 28.58 mm
Dimensions (L x W) 50.80 mm (W) x 25.40 mm (H)
Length (English) 10.94 yd.
Length (Metric) 10 m
Width (English) 2 in.
Width (Metric) 50.8 mm
Width (English) Printable Area 50.80 mm
Height (English) 1 in.
Height (Metric) 25.4 mm
Alternate Part Number THT-17-408-3