Learn More
Alfa Aesar™ Palladium, 3% on activated carbon powder, eggshell, unreduced, nominally 50% water wet
Marca:  Alfa Aesar™ 038295.14
Detalles adicionales : Peso : 0.02500kg
Cantidad. 25g
 Ver más versiones de este producto
Código de producto. 11314397
  • 266.00€ / 25 g
Stock EU 1
Añadir a la cesta
Especificaciones
7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03457879
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]
Especificaciones
Palladium
25g
7440-05-3
MFCD03457879
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
900 to 1100m2/g
Odorless
3% on activated carbon powder, Eggshell, unreduced, nominally 50% water wet
Pd
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
[Pd]
106.42
CHEBI:33363