missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK3 Recombinant Human Protein
PAK3 Recombinant Human Protein
Marca: Invitrogen PV3789
Detalles adicionales : Peso : 0.05250kg
Descripción
PAK proteins are critical effectors that link Rho GTPases to cytoskeleton reorganization and nuclear signaling. PAK proteins, a family of serine⁄threonine p21-activating kinases, serve as targets for the small GTP binding proteins Cdc42 and RAC and have been implicated in a wide range of biological activities.Especificaciones
NP_002569 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
68.6kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
PAK3, bPAK, hPAK3 | |
PAK3 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 100μM PAKtide peptide substrate (CRRKSLVGpTPYWMAPE) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
PAK3 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.43mg/mL | |
114294, 5063 | |
PAK3 Recombinant Human Protein | |
–80°C | |
1 Each | |
PAK3 Recombinant Human Protein | |
STE20 | |
150nmoles of phosphate transferred to PAKtide peptide substrate (CRRKSLVGpTPYWMAPE) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1.67μg/mL | |
His Tag | |
Kinase and inhibitors | |
90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.