missing translation for 'onlineSavingsMsg'
Learn More

PA28 Activator gamma Subunit/PSME3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Novus Biologicals NBP1-33498

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215740

  • 501.07€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

PA28 Activator gamma Subunit/PSME3 Polyclonal specifically detects PA28 Activator gamma Subunit/PSME3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.
Especificaciones

Especificaciones

PA28 Activator gamma Subunit/PSME3
Polyclonal
Unconjugated
0.1M Tris-Glycine (pH7), 10% Glycerol with 0.01% Thimerosal
11S regulator complex gamma subunit, 11S regulator complex subunit gamma, Activator of multicatalytic protease subunit 3, Ki, Ki antigen, PA28g, PA28gamma, PA28-gamma, PA28GKi nuclear autoantigen, proteasome (prosome, macropain) activator subunit 3 (PA28 gamma; Ki), Proteasome activator 28 subunit gamma, proteasome activator 28-gamma, proteasome activator complex subunit 3, REG-gamma
Rabbit
30 kDa
0.1 mL
Core ESC Like Genes, Immune System Diseases, Immunology, Neuroscience, Stem Cell Markers, Ubiquitin Proteasome Pathway
10197
Human, Mouse, Rat
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.65mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Knockout Validated
P61289
PSME3
Recombinant protein encompassing a sequence within the center region of human PSME3. The exact sequence is proprietary.
Affinity Purified
RUO
Primary
Tested in human and Mouse. Predicted cross-reactivity based on sequence identity: Pig (100%). Not yet tested in any other species.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only