missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p63 Mouse, Clone: TP63/1786, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 8626-MSM6-P1
Detalles adicionales : Peso : 0.01000kg
Especificaciones
p63 | |
Nuclear | |
TP63/1786 | |
Unconjugated | |
NP_001316893 | |
Mouse | |
63kDa | |
Protein A/G Purified | |
RUO | |
8626 | |
IgG2b κ |
Western Blot | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
TP63 | |
Recombinant human p63 protein fragment (aa3-106) | |
HEK293 cells or Prostate Carcinoma or Lung or bladder squamous cell carcinoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |