missing translation for 'onlineSavingsMsg'
Learn More

p504S/AMACR Mouse anti-Human, Mouse, Clone: 2H9B5, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 60240-1-IG-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817275

  • 319.00€ / 150 microlitros

Envío estimado: 21-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a racemase. The encoded enzyme interconverts pristanoyl-CoA and C27-bile acylCoAs between their (R)- and (S)-stereoisomers. The conversion to the (S)-stereoisomers is necessary for degradation of these substrates by peroxisomal beta-oxidation. Encoded proteins from this locus localize to both mitochondria and peroxisomes. Mutations in this gene may be associated with adult-onset sensorimotor neuropathy, pigmentary retinopathy, and adrenomyeloneuropathy due to defects in bile acid synthesis. Alternatively spliced transcript variants have been described.
Especificaciones

Especificaciones

p504S/AMACR
2H9B5
Unconjugated
AMACR
Liquid
Amacr
p504S,AMACR Fusion Protein Ag8720
Protein G
RUO
Antibody
Monoclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O09174, Q9UHK6
2 methylacyl CoA racemase, alpha methylacyl CoA racemase, AMACR, CBAS4, p504S, RACE
Mouse
IgG1
150 μL
-20°C
Primary
17117, 23600
Videos
SDS
Documentos
Certificados