Learn More
p35 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA564865
Detalles adicionales : Peso : 0.01000kg
Descripción
Immunogen sequence: FITPANVVFLY MLCRDVISSE VGSDHELQAV LLTCLYLSYS YMGNEISYPL KPFLVESCKE AFWDRCLSVI NLMSSKMLQI NADPHYFTQV FSDLKNESGQ EDK Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
p35 Polyclonal antibody specifically detects p35 in Human samples. It is validated for Immunocytochemistry.Especificaciones
p35 | |
Polyclonal | |
Unconjugated | |
Cdk5r1 | |
23 kDa subunit of tau-protein kinase II; cdk5 activator; CDK5 activator 1; CDK5P35; Cdk5r; Cdk5r1; cyclin dependent kinase 5 regulatory subunit 1; cyclin-dependent kinase 5 activator 1; Cyclin-dependent kinase 5 activator 1, p25; Cyclin-dependent kinase 5 activator 1, p35; Cyclin-dependent kinase 5 regulatory subunit 1; cyclin-dependent kinase 5, regulatory subunit (p35) 1; cyclin-dependent kinase 5, regulatory subunit 1 (p35); D11Bwg0379e; NCK5A; neuronal CDK5 activator; p23; p25; P25/P35; p35; p35nck5a; regulatory partner for CDK5 kinase; tau protein kinase II 23 kDa subunit; tau protein kinase II 23kDa subunit; TPKII regulatory subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8851 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15078 | |
Cdk5r1 | |
Recombinant Human p35 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only