Learn More
Velp Scientifica™ Glass Crucible P3
Product Code. 15309564 Shop All Velp Products
Change view
Click to view available options
For Use With (Equipment):
HU6 Hydrolysis Unit
Unit Size:
Pack of 6
Product Code. For Use With (Equipment) Unit Size
15309564 HU6 Hydrolysis Unit Pack of 6
1 options
This item is not returnable. View return policy
Product Code. 15309564 Supplier Velp Scientifica™ Supplier No. A00000087
  • ONLINE EXCLUSIVE PRICE
    359.00€
    332.00€ / paquete de 6
    Ahorro 27.00€
    7% Descuento
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Holds samples for general laboratory purposes. Velp Scientifica™ Glass Crucible P3 is an excellent accessory for Velp Scientifica™ FIWE Series Fiber Analyzers.
  • Durable glass
  • Standardized
Specifications
For Use With (Equipment) HU6 Hydrolysis Unit
Product Type Crucible P3
Material Glass
Quantity 6 Pack