missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonylacetic acid, 98%

Product Code. 11301517
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11301517 25 g 25g
11391507 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11301517 Supplier Thermo Scientific Alfa Aesar Supplier No. L02102.14
 Restricted Item
Fecha estimada de envoi: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3937-96-0 | C9H10O4S | 214.235 g/mol

p-Toluenesulfonylacetic acid is used as pharmaceuticals intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluenesulfonylacetic acid is used as pharmaceuticals intermediates.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3937-96-0
Molecular Formula C9H10O4S
Molecular Weight (g/mol) 214.235
MDL Number MFCD00021764
InChI Key AQDHXMBUTDLAMD-UHFFFAOYSA-N
Synonym 2-tosylacetic acid, 4-toluenesulfonylacetic acid, 4-methylphenyl sulfonyl acetic acid, 2-p-toluenesulfonyl acetic acid, tosylacetic acid, unii-yid78nf77o, p-toluenesulphonylacetic acid, 4-toluenesulphonylacetic acid, p-toluenesulfonylacetic acid, yid78nf77o
PubChem CID 77538
IUPAC Name 2-(4-methylphenyl)sulfonylacetic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)CC(=O)O

Specifications

Melting Point 117°C to 119°C
Quantity 25 g
Beilstein 2331311
Solubility Information Insoluble in water.
Formula Weight 214.24
Percent Purity 98%
Chemical Name or Material p-Toluenesulfonylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.