Learn More
Oxymetazoline Hydrochloride, LoGiCal
Product Code. 30458691
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30458691 10 mg 10mg
1 options
This item is not returnable. View return policy
Product Code. 30458691 Supplier LoGiCal Supplier No. LGCFOR0177.00
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Oxymetazoline Hydrochloride
CAS 2-8-2315
Analyte or Component Names Oxymetazoline Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C16 H24 N2 O . Cl H
Certifications/Compliance ISO 9001
Synonym Oxymetazoline Hydrochloride, 3-[(4,5-Dihydro-1H-imidazol-2-yl)methyl]-6-(1,1-dimethylethyl)-2,4-dimethylphenol hydrochloride (1:1), 3-[(4,5-Dihydro-1H-imidazol-2-yl)methyl]-6-(1,1-dimethylethyl)-2,4-dimethylphenol monohydrochloride, 6-tert-Butyl-3-(2-imidazolin-2-ylmethyl)-2,4-dimethylphenol hydrochloride, 6-tert-Butyl-3-(2-imidazolin-2-ylmethyl)-2,4-dimethylphenol monohydrochloride, 2,6-Dimethyl-4-tertiarybutyl-3-hydroxyphenyl)methylimidazoline hydrochloride, 2-(4-tert-Butyl-2,6-dimethyl-3-hydroxybenzyl)-2-imidazolinium chloride, 4-Way Nasal 12 Hour Spray, 4-Way Nasal Spray, 6-tert-Butyl-3-(2-imidazolin-2-ylmethyl)-2,4-dimethylphenol hydrochloride, Afrazine, Afrin, Afrin hydrochloride, Allerest 12 Hour Nasal Spray, Dristan Long-Lasting Nasal Mist, Duration, Duration 12 Hour Nasal Spray, H 990, Iliadin, Nafrine, Nasivin, Neo-Synephrine 12 Hour, Neo-Synephrine 12 Hour NTZ, Nostrilla, Ocuclear, Oxilin, Oxymetazoline chloride, Oxymetazoline hydrochloride, Respir, Sch 9384
InChI Formula InChI=1S/C16H24N2O.ClH/c1-10-8-13(16(3,4)5)15(19)11(2)12(10)9-14-17-6-7-18-14;/h8,19H,6-7,9H2,1-5H3,(H,17,18);1H
Solution Type Neat
SMILES Cl.Cc1cc(c(O)c(C)c1CC2=NCCN2)C(C)(C)C
IUPAC Name 6-tert-butyl-3-(4,5-dihydro-1H-imidazol-2-ylmethyl)-2,4-dimethylphenol;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 296.84
Quantity 10 mg
Formula Weight 296.1655 g/mol