Learn More
Thermo Scientific™ Oxacillin M.I.C.Evaluator Strips (M.I.C.E.™)
Product Code. 11454039
Change view
Click to view available options
Description:
Oxacillin M.I.C.Evaluator Strips (M.I.C.E.™)
Concentration:
256-0.015µg/mL
Unit Size:
Pack of 10
Product Code. Description Concentration Unit Size
11454039 Oxacillin M.I.C.Evaluator Strips (M.I.C.E.™) 256-0.015µg/mL Pack of 10
1 options
This item is not returnable. View return policy
Product Code. 11454039 Supplier Thermo Scientific™ Supplier No. MA0114D
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Determine accurate Minimum Inhibitory Concentration (MIC) values of antibiotics using MIC testing strips.
Effortlessly establish accurate Minimum Inhibitory Concentration (MIC) values manually using Thermo Scientific™ Oxoid™ Oxacillin M.I.C.Evaluator Strips (M.I.C.E.™).
Conveniently packaged M.I.C.E. strips combine the ease of a diffusion test with the accuracy of an MIC test.
Simple to Perform
  • Distinctive gradient format provides excellent contrast with the agar
  • Increased font size makes reading easier
  • Scale conforms to international standards
  • Strips are tailored to meet clinical requirements while maintaining the accuracy required for research
Easy-to-handle
  • The strip is conveniently presented when the sachet is peeled apart
  • Single-strip packaging eliminates the requirement to maintain and quality control opened strips; open one and use it
  • Stability of the individual M.I.C.E. strips is ensured through the inclusion of a desiccant within each sachet
  • 10 and 50 pack sizes offer users maximum flexibility to put together custom panels for testing
  • Easily stackable boxes simplify storage; all antimicrobials are stored in the fridge (2-8°C)
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Spezifikation
Concentration 256-0.015µg/mL
Content And Storage 2°C to 8°C
Description Oxacillin M.I.C.Evaluator Strips (M.I.C.E.™)
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.