missing translation for 'onlineSavingsMsg'
Learn More

Oval Cell Marker Antibody (OC2-3C5), Alexa Fluor™ 350, Novus Biologicals™

Rat Monoclonal Antibody

Marca:  Bio-Techne NBP1-18968AF350

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216051

  • 561.54€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

Oval Cell Marker Monoclonal specifically detects Oval Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Especificaciones

Especificaciones

Oval Cell Marker
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
Nonparenchymal cells from DDC-treated mice to induce the oval cell response.
Primary
Mouse
IgG2a
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
OC2-3C5
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Rat
0.1 mL
Specific for both proliferating and normal hepatic periductal and ductal cells.
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales