missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSTM1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14621-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein that may be involved in the degradation of G proteins via the ubiquitin-dependent proteasome pathway. The encoded protein binds to members of subfamily A of the regulator of the G-protein signaling (RGS) family through an N-terminal leucine-rich region. This protein also has a central RING finger-like domain and E3 ubiquitin ligase activity. This protein is highly conserved from flies to humans. Defects in this gene may cause the autosomal recessive, infantile maligt form of osteopetrosis.Especificaciones
OSTM1 | |
Polyclonal | |
Unconjugated | |
OSTM1 | |
GIPN, GL, HSPC019, OPTB5, OSTM1, UNQ6098/PRO21201 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14628, 28962 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q86WC4, Q8BGT0 | |
OSTM1 | |
OSTM1 Fusion Protein Ag6254 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |