Learn More
ORM2-Specific Mouse, anti-Human, CoraLite 488, Clone: 4E9H10, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech CL488-66217-100UL
Detalles adicionales : Peso : 0.00020kg
Código de producto. 17231556
  • 367.00€ / 100 microlitros
Fecha estimada de envío: 13-05-2024
para ver el stock
Descripción
Descripción
ORM2, also named as AGP2 and OMD 2, belongs to the calycin superfamily and Lipocalin family. It functions as transport protein in the blood stream. ORM2 binds various hydrophobic ligands in the interior of its beta-barrel domain. It also binds synthetic drugs and influences their distribution and availability. ORM2 appears to function in modulating the activity of the immune system during the acute-phase reaction. ORM2 is one of the conserved endoplasmic reticulum membrane proteins which regulating lipid homeostasis and protein quality control. This antibody can specially recognize ORM2. It recognizes a band about 45 kDa in human plasma which may be due to the glycosylation of ORM2 or the dimer formation of the protein.
Especificaciones
Especificaciones
ORM2-Specific
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Immunofluorescence
4E9H10
CoraLite 488
100 μL
5005