Learn More
ORM2-Specific Mouse, anti-Human, CoraLite 488, Clone: 4E9H10, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-66217-100UL
Detalles adicionales : Peso : 0.00020kg
Descripción
ORM2, also named as AGP2 and OMD 2, belongs to the calycin superfamily and Lipocalin family. It functions as transport protein in the blood stream. ORM2 binds various hydrophobic ligands in the interior of its beta-barrel domain. It also binds synthetic drugs and influences their distribution and availability. ORM2 appears to function in modulating the activity of the immune system during the acute-phase reaction. ORM2 is one of the conserved endoplasmic reticulum membrane proteins which regulating lipid homeostasis and protein quality control. This antibody can specially recognize ORM2. It recognizes a band about 45 kDa in human plasma which may be due to the glycosylation of ORM2 or the dimer formation of the protein.
Especificaciones
ORM2-Specific | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Immunofluorescence | |
4E9H10 | |
CoraLite 488 | |
100 μL | |
5005 | |
Liquid |