Learn More
Thermo Scientific™ Orion™ Star A Series Portable Meter Protective Armor
Product Code. 11631489
Change view
Click to view available options
Star A Protector Armor
Product Code.
11631489 Star A Protector Armor
1 options
This item is not returnable. View return policy
Product Code. 11631489 Supplier Thermo Scientific™ Supplier No. STARAAR
  • ONLINE EXCLUSIVE PRICE
    170.95€
    139.23€ / paquete de 1
    Ahorro 31.72€
    18% Descuento
Fecha estimada de envoi: 29-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Protect your portable meter using Thermo Scientific™ Orion™ Star A Series Portable Meter Protective Armor for use with Thermo Scientific™ Orion™ Star A series portable meters
Protect your portable meter using Thermo Scientific™ Orion™ Star A Series Portable Meter Protective Armor for use with Thermo Scientific™ Orion™ Star A series portable meters (Star A320, Star A220 and Star A120 series). Portable meter armor adds protection around the meter screen and includes a hand or belt strap that can be converted to built-in stand, pH electrode holder, conductivity and DO probe holder and pH electrode storage sleeve.
TRUSTED_SUSTAINABILITY
Specifications
Description Star A Protector Armor
Type Meter Accessories
Includes Orion Star A series portable meter protective armor with hand strap and built-in stand, 810017 pH electrode storage sleeve, STARA-ESPH pH electrode holder, STARA-ESCD conductivity and dissolved oxygen sensor holder
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.