Learn More
Thermo Scientific™ Orion™ Filling Solutions for Process Analyzer Reference Electrodes
Product Code. 15827134
Change view
Click to view available options
For Use With (Equipment):
100056/210056 Reference Electrodes
100057 Reference Electrode
Unit Size:
480mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15827134 100056/210056 Reference Electrodes Each
16815623 100057 Reference Electrode 480mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15827134 Supplier Thermo Scientific™ Supplier No. 181073
 Restricted Item
Fecha estimada de envoi: 07-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Enhance the performance and extend the life of ion selective electrodes used in our process analyzers with this filling solution.
Enhance the performance and extend the life of ion selective electrodes used in Orion™ process analyzers with Thermo Scientific™ Orion™ Filling Solutions for Process Analyzer Reference Electrodes.
Filling solutions for Orion process analyzer reference electrodes
TRUSTED_SUSTAINABILITY
Specifications
Type Filling Solution
Quantity 5 x 60 mL
For Use With (Equipment) 100056/210056 Reference Electrodes
compliance-icons
Product Identifier
  • Reference Electrode Internal Filling Solution
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: cesium chloride, triton x-100
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.