Learn More
Hirschmann™ Opus™ Dispenser Hand Switch
Product Code. 10401541
Change view
Click to view available options
Type:
Hand Key
Unit Size:
Each
Product Code. Type Unit Size
10401541 Hand Key Each
1 options
This item is not returnable. View return policy
Product Code. 10401541 Supplier Hirschmann™ Supplier No. 9564001
  • ONLINE EXCLUSIVE PRICE
    109.20€
    71.74€ / paquete de 1
    Ahorro 37.46€
    34% Descuento
Fecha estimada de envoi: 01-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Hand switch for remotely triggering the dispensing step with an opus™ dispenser.
  • For opus™
  • Programmable
Specifications
For Use With (Application) Starting the Dosing Operation (For Basic Units 10 + 20 + 50 mL)
No. per Pack 1 Each
Type Hand Key