missing translation for 'onlineSavingsMsg'
Learn More

Opticin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 17897-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817904

  • 109.00€ / 20 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Opticin, also known as OPTC or OPT, is a 332 amino acid protein that is secreted into the extracellular matrix and contains seven LRR repeats. Expressed in skin and fetal liver and also present in ocular tissues, such as the retina, cornea and iris, opticin functions to bind collagen fibrils and, via this interaction, is thought to regulate fibril spacing, morphology and organization. Opticin is subject to post-translational O-glycosylation and may be associated with the pathogenesis of primary open angle glaucoma.
Especificaciones

Especificaciones

Opticin
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q920A0, Q9UBM4
Oculoglycan, OPT, OPTC, Opticin
Rabbit
IgG
20 μL
-20°C
Primary
26254, 269120, 304802
Western Blot
Unconjugated
OPTC
Liquid
Optc
Opticin Fusion Protein Ag12257
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human, Mouse
Videos
SDS
Documentos
Certificados