Learn More
Online Internal Standard Kits
Product Code. 17805296 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Online Internal Standard Kit
For Use With (Equipment):
iCAP MX and iCAP Q/Qnova ICP-MS Systems
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Unit Size
17805296 Online Internal Standard Kit iCAP MX and iCAP Q/Qnova ICP-MS Systems Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17805296 Supplier Thermo Scientific Supplier No. 1324690
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Get a reliable solution for accurate sample dilution with the Thermo Scientific™ Online Internal Standard Kit for our Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems.
Take advantage of our Online Internal Standard Kit for iCAP MX and iCAP Q/Qnova Series ICP-MS systems. This kit provides a reliable solution for accurate sample dilution, an essential step in ICP-MS analysis. It includes high-quality parts designed for precise and consistent dilution, ensuring accurate and reproducible results.
Ensure accurate and reproducible results
  • Reliable solution for accurate online sample dilution in ICP-MS analysis
  • Includes high-quality parts designed for precise and consistent online internal standard addition
Specifications
Type Online Internal Standard Kit
For Use With (Equipment) iCAP MX and iCAP Q/Qnova ICP-MS Systems