Learn More
Omnifit™ HiT Chromatography Columns
Omnifit™ HiT Chromatography Columns withstand temperatures up to 302°F (150°C)—use with heating block or jacket
Marca:  Kinesis 006HCC-06-15-AF
Detalles adicionales : Peso : 0.34000kg
 Ver más versiones de este producto
Código de producto. 15845554
  • 474.00€ / cada uno
Fecha de envío estimada
del proveedor 29-10-2018
Añadir a la cesta
Includes: Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end and a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits).
Descripción
Descripción
  • Adjustable endpiece design for quick and easy height adjustment
  • Ideal for both in batch production or within a flow chemistry systems
  • Reusable columns let you pack your own media specific to your application
Especificaciones
Especificaciones
Omnifit™ HiT™ Chromatography Column w/ 1 Fixed & 1 Adjustable Endpiece, 6.6 x 150mm; 1/ea
Liquid Chromatography (LC)
(1) fixed and (1) adjustable
10 μm and 30 μm
150mm
150°C
4-150°C
6.6mm
900 psi
1.4 to 4.1mL
Borosilicate glass, PEEK, FKM/FPM, PTFE
Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end and a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits).
Videos