Learn More
Novus Biologicals™ Olfactomedin-2/Noelin-2 Overexpression Lysate
Marca:  Novus Biologicals™ NBL1-13926
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214494
  • 685.73€ / 0.10 miligramo
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
The Olfactomedin-2/Noelin-2 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Especificaciones
Especificaciones
Western Blot
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
The lysate was created in HEK293T cells, using plasmid ID RC23786 and based on accession number NM_58164. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens olfactomedin 2 (OLFM2), mRNA.
Native