Learn More
Fisherbrand™ Pump Oil
Product Code. 11767929 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Volume (Metric):
10 L
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
11767929 10 L Each
1 options
This item is not returnable. View return policy
Product Code. 11767929
  • ONLINE EXCLUSIVE PRICE
    411.00€
    302.41€ / paquete de 1
    Ahorro 108.59€
    26% Descuento
 Restricted Item
Fecha estimada de envoi: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Lubricate pumps with this oil. Fisherbrand™ Pump Oil is designed for use with Fisherbrand™ rotary vane pumps.
  • Generic mineral oil for use in most one and two stage rotary vane pumps
  • 20L bottle
  • Low vapor pressure
  • For general use in rotary vane pumps
TRUSTED_SUSTAINABILITY
Specifications
Product Type Oil
For Use With (Equipment) Rotary Vane Pump
Volume (Metric) 10 L