missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18996-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The antibody is specific to OCT2.
OCT-2 is a transcription factor belonging to the POU homeo-domain family that binds to the Ig gene octamer sites regulating B cell specific genes. OCT-2 protein expression is not restricted to B cells, although expression levels are much higher in these cells. OCT-2 expression is reported to be significantly greater in germinal center derived lymphomas, although other B cell lymphomas also display high levels of expression.Especificaciones
OCT2 | |
Polyclonal | |
Unconjugated | |
POU2F2 | |
OCT2, Octamer binding protein 2, OTF 2, OTF2, POU class 2 homeobox 2, POU2F2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117058, 18987, 5452 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P09086, Q00196 | |
Pou2f2 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |