Learn More
Clorhidrato de o-tolilhidracina, 98 %, Thermo Scientific Chemicals
Catalog Number 11442428
Click to view available options
Cantidad:
5 g
25 g
100 g
635-26-7
C7H11ClN2
158.629
MFCD00012925
KJGFNDCSTWGUDT-UHFFFAOYSA-N
o-tolylhydrazine hydrochloride, 2-methylphenyl hydrazine hydrochloride, 2-methylphenylhydrazine hydrochloride, o-tolylhydrazine hcl, 1-2-methylphenyl hydrazine hydrochloride, 2-tolylhydrazine hydrochloride, o-tolylhydrazinium 1+ chloride, o-methylphenylhydrazine hcl, 2-methylphenylhydrazine hcl, hydrazine, 2-methylphenyl-, monohydrochloride
71554
(2-metilfenil)hidracina; clorhidrato
CC1=CC=CC=C1NN.Cl
635-26-7
C7H11ClN2
158.629
MFCD00012925
KJGFNDCSTWGUDT-UHFFFAOYSA-N
o-tolylhydrazine hydrochloride, 2-methylphenyl hydrazine hydrochloride, 2-methylphenylhydrazine hydrochloride, o-tolylhydrazine hcl, 1-2-methylphenyl hydrazine hydrochloride, 2-tolylhydrazine hydrochloride, o-tolylhydrazinium 1+ chloride, o-methylphenylhydrazine hcl, 2-methylphenylhydrazine hcl, hydrazine, 2-methylphenyl-, monohydrochloride
71554
(2-metilfenil)hidracina; clorhidrato
CC1=CC=CC=C1NN.Cl
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material o-Tolylhydrazine hydrochloride
Punto de fusión ∼190°C (decomposition)
Cantidad 25 g
Número UN UN2811
Beilstein 3655929
Sensibilidad Hygroscopic
Formula Weight (peso de la fórmula) 158.63
Porcentaje de pureza 98%