Learn More
O-Desethyl Dapagliflozin (>80%), TRC
Product Code. 30005016
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005016 25 mg 25mg
30011198 5 mg 5mg
30005027 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005016 Supplier Toronto Research Chemicals Supplier No. TRCD29054325MG
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 864070-37-1
Molecular Formula C19 H21 Cl O6
Molecular Weight (g/mol) 380.82
Synonym D-Glucitol, 1,5-anhydro-1-C-[4-chloro-3-[(4-hydroxyphenyl)methyl]phenyl]-, (1S)-, (1S)-1,5-Anhydro-1-C-[4-chloro-3-[(4-hydroxyphenyl)methyl]phenyl]-D-glucitol, (2S,3R,4R,5S,6R)-2-[4-chloro-3-[(4-hydroxyphenyl)methyl]phenyl]-6-(hydroxymethyl)tetrahydropyran-3,4,5-triol
IUPAC Name (2S,3R,4R,5S,6R)-2-[4-chloro-3-[(4-hydroxyphenyl)methyl]phenyl]-6-(hydroxymethyl)tetrahydropyran-3,4,5-triol
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2ccc(Cl)c(Cc3ccc(O)cc3)c2
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C19H21ClO6/c20-14-6-3-11(8-12(14)7-10-1-4-13(22)5-2-10)19-18(25)17(24)16(23)15(9-21)26-19/h1-6,8,15-19,21-25H,7,9H2/t15-,16-,17+,18-,19+/m1/s1
Formula Weight 380.1027
Percent Purity >95%
Chemical Name or Material Empagliflozin Impurity 3
Grade HPLC