Learn More
Thermo Scientific™ Nunc™ Containers
BIOPHARMA
Product Code. 10793133
Change view
Click to view available options
Capacity (Metric):
12.5 mL
20 mL
30 mL
50 mL
Unit Size:
Pack of 1000
Pack of 1200
Pack of 2400
Pack of 800
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10793133 20 mL Pack of 2400
10578531 12.5 mL Pack of 1000
11301765 30 mL Pack of 1200
10428892 50 mL Pack of 800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10793133 Supplier Thermo Scientific™ Supplier No. 536323
  • ONLINE EXCLUSIVE PRICE
    725.00€
    668.00€ / paquete de 2400
    Ahorro 57.00€
    7% Descuento
View all promotions
En stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Store samples safely using Thermo Scientific™ Nunc™ Containers.
Store samples safely using Thermo Scientific™ Nunc™ Containers. Nunc Labware Products are made from high purity resins and molded using our state-of-the-art processes. Plastic labware is a safer alternative to glass without sacrificing accuracy.
Features:
  • Transparent polystyrene construction
  • Versatile containers made of high-quality resins
  • Snap caps available for all containers (not included)
  • Nonsterile (except for 561301)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Container
Closure Included No
Material Polystyrene
Shape Cylindrical
Sterility Non-sterile
Capacity (Metric) 20 mL
Closure Type Snap Cap
Color Clear