missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15731-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is one of several human genes similar to the C. elegans gene Fox-1. This gene encodes an RNA binding protein that is thought to be a key regulator of alternative exon splicing in the nervous system and other cell types. The protein binds to a conserved UGCAUG element found downstream of many alternatively spliced exons and promotes inclusion of the alternative exon in mature transcripts. The protein also interacts with the estrogen receptor 1 transcription factor and regulates estrogen receptor 1 transcriptional activity. Multiple transcript variants encoding different isoforms have been found for this gene.Especificaciones
NUF2 | |
Polyclonal | |
Unconjugated | |
NUF2 | |
CDCA1, hNuf2, hNuf2R, hsNuf2, Kinetochore protein Nuf2, NUF2, NUF2R | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
304951, 66977, 83540 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6AYL9, Q99P69, Q9BZD4 | |
Nuf2 | |
NUF2 Fusion Protein Ag8479 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |