missing translation for 'onlineSavingsMsg'
Learn More

NUDT12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 17487-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817384

  • 109.00€ / 20 microlitros

Envío estimado: 28-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.
Especificaciones

Especificaciones

NUDT12
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BQG2, Q9DCN1
Nudix motif 12, NUDT12
Rabbit
IgG
20 μL
-20°C
Primary
367323, 67993, 83594
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
NUDT12
Liquid
Nudt12
NUDT12 Fusion Protein Ag11557
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Rat, Human
Videos
SDS
Documentos
Certificados