missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleophosmin (Acute Myeloid Leukemia Marker) Mouse, Clone: NA24, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 4869-MSM3-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Nucleophosmin (Acute Myeloid Leukemia Marker) | |
Nuclear and Cytoplasmic | |
NA24 | |
Unconjugated | |
Mouse | |
33kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
4869 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
GST fusion protein containing the N-terminal part of nucleophosmin fused to 14 amino acid of ALK protein | |
MCF-7, HeLa or A431 cells. Skin, Colon | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |