Learn More
Macherey-Nagel™ NucleoMag X32 - System For Automated DNA/RNA Purificat
Beskrivelse
- Capable of processing up to 32 samples simultaneously, the stand-alone instrument is easy to set up, program, and operate
- Mixing, magnetic bead transfer, washing, and elution steps are performed automatically, saving valuable hands-on time
- The purification process can be completed within 20 to 60 minutes depending on the type of NucleoMag kit used and the complexity of the program
Tekniske data
Tekniske data
| Product Type | Nucleic Acid Extraction System |
| Sample Type | Tissue |
| Suitable Downstream Analysis | PCR/qPCR, Sequencing, Mass Spec |
| Volume (Metric) Processing | 50 to 1000 μL |
| Plates per Deck | 4 |
| Format | 96-well Plate |
| Capacity | 1 to 32 Samples |
| Content And Storage | NucleoMag X32 Automated DNA/RNA Purification System, Power Cable, Manual |
| For Use With (Application) | Automated Extraction System For Magnetic Bead-based DNA/RNA Purification |
| Product Line | NucleoMag |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?