missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR2F6 Mouse anti-Human, Clone: 2H2B8, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60117-2-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Transcription factor predomitly involved in transcriptional repression. Binds to promoter/enhancer response elements that contain the imperfect 5′-AGGTCA-3′ direct or inverted repeats with various spacings which are also recognized by other nuclear hormone receptors. Involved in modulation of hormonal responses. Represses transcriptional activity of the lutropin-choriogonadotropic hormone receptor/LHCGR gene, the renin/REN gene and the oxytocin-neurophysin/OXT gene. Represses the triiodothyronine-dependent and -independent transcriptional activity of the thyroid hormone receptor gene in a cell type-specific manner. The corepressing function towards thyroid hormone receptor beta/THRB involves at least in part the inhibition of THRB binding to triiodothyronine response elements (TREs) by NR2F6. Inhibits NFATC transcription factor DNA binding and subsequently its transcriptional activity. Acts as transcriptional repressor of IL-17 expression in Th-17 differentiated CD4(+) T cells and may be involved in induction and/or maintece of peripheral immunological tolerance and autoimmunity. Involved in development of forebrain circadian clock; is required early in the development of the locus coeruleus (LC).Especificaciones
NR2F6 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P10588 | |
NR2F6 | |
NR2F6 Fusion Protein Ag7614 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
2H2B8 | |
Unconjugated | |
NR2F6 | |
EAR 2, EAR2, ERBAL2, NR2F6, V erbA related protein 2 | |
Mouse | |
Protein G | |
RUO | |
2063 | |
-20°C | |
Liquid |