missing translation for 'onlineSavingsMsg'
Learn More

NOX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 19013-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818834

  • 109.00€ / 20 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Mox1 and the glycoprotein gp91-phox are largely related proteins that are essential components of the NADPH oxidase. The superoxide-generating NADPH oxidase complex expresses in phagocytes, neuroepithelial bodies, vascular smooth muscle cells, and endothelial cells. It includes a membranebound flavocytochrome containing two subunits, gp91-phox and p22-phox, and the cytosolic proteins p47-phox and p67-phox. The p22- and gp91-phox subunits also function as surface O2 sensors that initiate cellular signaling in response to hypoxic conditions. Mox1 and gp91 contain identical C-terminal sequence identity, yet possess distinct expression patterns. gp91-phox expresses in eosinophils, neutrophils, monocytes, and B-lymphocytes, whereas Mox1 is predomitly detected in the colon, with low expression in the uterus and prostate.
Especificaciones

Especificaciones

NOX2
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P04839, Q61093
CYBB, Cytochrome b 245 heavy chain, NOX2, p91 PHOX
Rabbit
IgG
20 μL
-20°C
Primary
13058, 1536, 66021
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CYBB
Liquid
Cybb
NOX2 Fusion Protein Ag5536
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human, Mouse
Videos
SDS
Documentos
Certificados